missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD45R (B220) Rat anti-Human, Mouse, FITC, Clone: RA3-6B2, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS11045281

 View more versions of this product

Catalog No. 501129639



Description

Description

Description: The RA3-6B2 monoclonal antibody reacts with exon A-restricted isoform of mouse CD45, a 220 kDa surface molecule. CD45R/B220 epitope is mainly expressed by the B cell lineage from early Pro-B to mature B cells. However, some activated T cells, lymphokine activated killer cells (LAK), NK cell progenitors in the bone marrow, and T cells of the lpr/lpr mutant mouse also express this antigen. Applications Reported: This RA3-6B2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RA3-6B2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD45R is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. CD45R contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains. It is specifically expressed in hematopoietic cells and has been shown to be an essential regulator of T- and B-cell antigen receptor signaling.
Specifications

Specifications

CD45R (B220)
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P06800, P08575
B220, CD45R, Cd45, Ly-5, Lyt-4, T200, Cluster designation 45R
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Flow Cytometry
RA3-6B2
FITC
PTPRC
Liquid
Ptprc
IgG2a, kappa
50 μg
4° C, store in dark, DO NOT FREEZE!
Primary
19264, 5788
Documents

For Research Use Only.