missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD45R (B220) Rat anti-Human, Mouse, FITC, Clone: RA3-6B2, eBioscience™

Rat Monoclonal Antibody

$56.55 - $723.55

Specifications

Antigen CD45R (B220)
Clone RA3-6B2
Host Species Rat
Gene Alias Ly-5, Lyt-4, T200
Species Reactivity Human, Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509573 Life Technologies
LS11045263
10μg Each for $56.55
509576 Life Technologies
LS11045285
500μg Each for $390.05
509577 Life Technologies
LS11045286
1mg Each for $723.55
501129639 Life Technologies
LS11045281
50μg Each for $71.05
501129688 Life Technologies
LS11045282
100μg Each for $114.55
Specifications

Description

The RA3-6B2 monoclonal antibody reacts with exon A-restricted isoform of mouse CD45, a 220kDa surface molecule. CD45R/B220 epitope is mainly expressed by the B cell lineage from early Pro-B to mature B cells. However, some activated T cells, lymphokine activated killer cells (LAK), NK cell progenitors in the bone marrow, and T cells of the lpr/lpr mutant mouse also express this antigen.
Description & Specifications

Specifications

CD45R (B220)
Rat
Human, Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
RA3-6B2
Ly-5, Lyt-4, T200
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL