Please login to your online account to display your discounted pricing

CD45R (B220) Rat anti-Human, Mouse, eVolve 605, Clone: RA3-6B2, eBioscience™

Rat Monoclonal Antibody

$203.00 - $423.40

Specifications

Antigen CD45R (B220)
Clone RA3-6B2
Host Species Rat
Gene Alias Ly-5, Lyt-4, T200
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125015

 
life technologies
LS83045241
25 Tests Each for $203.00

501125016

 
life technologies
LS83045242
100 Tests Each for $423.40
Description & Specifications

Specifications

Antigen CD45R (B220)
Clone RA3-6B2
Host Species Rat
Gene Alias Ly-5, Lyt-4, T200
Species Reactivity Human, Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eVolve 605
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL

The RA3-6B2 monoclonal antibody reacts with exon A-restricted isoform of mouse CD45, a 220kDa surface molecule. CD45R/B220 epitope is mainly expressed by the B cell lineage from early Pro-B to mature B cells. However, some activated T cells, lymphokine activated killer cells (LAK), NK cell progenitors in the bone marrow, and T cells of the lpr/lpr mutant mouse also express this antigen.