Please login to your online account to display your discounted pricing

CD45R (B220) Rat anti-Human, Mouse, Clone: RA3-6B2, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$201.55 - $651.05

Specifications

Antigen CD45R (B220)
Clone RA3-6B2
Host Species Rat
Gene Alias Ly-5, Lyt-4, T200
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014321

 
life technologies
LS16045282
100μg Each for $201.55

5014322

 
life technologies
LS16045285
500μg Each for $419.05

5014323

 
life technologies
LS16045286
1mg Each for $651.05
Description & Specifications

Specifications

Antigen CD45R (B220)
Clone RA3-6B2
Host Species Rat
Gene Alias Ly-5, Lyt-4, T200
Species Reactivity Human, Mouse
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The RA3-6B2 monoclonal antibody reacts with exon A-restricted isoform of mouse CD45, a 220kDa surface molecule. CD45R/B220 epitope is mainly expressed by the B cell lineage from early Pro-B to mature B cells. However, some activated T cells, lymphokine activated killer cells (LAK), NK cell progenitors in the bone marrow, and T cells of the lpr/lpr mutant mouse also express this antigen.