missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD45R (B220) Rat anti-Human, Mouse, Biotin, Clone: RA3-6B2, eBioscience™

Rat Monoclonal Antibody

$71.05 - $709.05

Specifications

Antigen CD45R (B220)
Clone RA3-6B2
Host Species Rat
Gene Alias Ly-5, Lyt-4, T200
Species Reactivity Human, Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011551 Life Technologies
LS13045281
50μg Each for $71.05
5011552 Life Technologies
LS13045282
100μg Each for $100.05
5011553 Life Technologies
LS13045285
500μg Each for $361.05
5011554 Life Technologies
LS13045286
1mg Each for $709.05
Specifications

Description

The RA3-6B2 monoclonal antibody reacts with exon A-restricted isoform of mouse CD45, a 220kDa surface molecule. CD45R/B220 epitope is mainly expressed by the B cell lineage from early Pro-B to mature B cells. However, some activated T cells, lymphokine activated killer cells (LAK), NK cell progenitors in the bone marrow, and T cells of the lpr/lpr mutant mouse also express this antigen.
Description & Specifications

Specifications

CD45R (B220)
Rat
Human, Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
RA3-6B2
Ly-5, Lyt-4, T200
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL