Please login to your online account to display your discounted pricing

CD45R (B220) Rat anti-Canine, PE, Clone: YKIX753.22, eBioscience™

Rat Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen CD45R (B220)
Clone YKIX753.22
Host Species Rat
Species Reactivity Canine
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010828

 
affymetrix
12545141
25 tests Each for $143.55

5010829

 
affymetrix
12545142
100 tests Each for $361.05
Description & Specifications

Specifications

Antigen CD45R (B220)
Clone YKIX753.22
Host Species Rat
Species Reactivity Canine
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.0075μg)/test

The YKIX753.22 monoclonal antibody reacts with canine CD45R. In dogs, CD45R is expressed on B cells and peripheral T cells. In addition, CD45 exists as multiple isoforms due to alternative splicing. Some isoforms of CD45 are CD45RA, CD45RB, or CD45RO depending on the exon used during transcription. The C-terminal domain of CD45 exhibits tyrosine phosphatase activity that enables signaling from by T cell and B cell receptors following activation.

This antibody has been reported to promote cell depletion via antibody-dependent cell-mediated cytotoxicity when used in conjunction with YKIX716.13.