Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD45/T200/LCA Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, and flow cytometry experiments.

$258.47 - $723.88

Specifications

Antigen CD45/T200/LCA Ab-2
Clone PD7/26/16 + 2B11
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS355P

 
thermo scientific™ lab vision™
MS355P
Purified Ab with BSA and Azide 1mL Each for $667.45

MS355P0

 
thermo scientific™ lab vision™
MS355P0
Purified Ab with BSA and Azide 100μL Each for $258.47

MS355P1

 
thermo scientific™ lab vision™
MS355P1
Purified Ab with BSA and Azide 500μL Each for $510.86

MS355P1ABX

 
thermo scientific™ lab vision™
MS355P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $584.90

MS355PABX

 
thermo scientific™ lab vision™
MS355PABX
Ab without BSA/Azide (Frozen) 200μL Each for $723.88

MS355R7

 
thermo scientific™ lab vision™
MS355R7
Ready-to-use for IHC (UVLP) 7mL Each for $426.08
Description & Specifications

Specifications

Antigen CD45/T200/LCA Ab-2
Clone PD7/26/16 + 2B11
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Host Species Murine
Regulatory Status IVD
Research Discipline Oncogenesis
Type Primary
Immunogen PD7/26/16: human peripheral blood lymphocytes maintained in T cell growth factor. 2B11: isolated neoplastic cells from T cell lymphoma.

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, and flow cytometry experiments with Thermo Scientific™ CD45/T200/LCA Ab-2, Mouse Monoclonal Antibody.

CD45 leucocyte common antigen (LCA) belongs to the family of at least four isoforms of membrane glycoproteins (220, 205, 190, 180 kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and malignant non-hematopoietic tissues. The intracellular portion of these molecules have protein phosphatase activity and are involved in regulation of transmembrane signals. Antibody to CD45 is useful in differential identification of lymphoid tumors from non-hematopoietic undifferentiated neoplasms.

Host Species: Mouse

Clone: PD7/26/16 + 2B11

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat and dog. Others not known

Epitope: Not determined

Immunogen: PD7/26/16: human peripheral blood lymphocytes maintained in T-cell growth factor. 2B11: isolated neoplastic cells from T-cell lymphoma

Molecular Weight: 220, 205, 190, 180 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry