Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD45/T200/LCA Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD45/T200/LCA Ab-1, Mouse Monoclonal Antibody.

$238.04 - $687.49

Specifications

Antigen CD45/T200/LCA Ab-1
Clone Bra55
Applications Flow Cytometry
Applications Immunofluorescence
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS240P

 
thermo scientific™ lab vision™
MS240P
200μg/mL 1mL Each for $603.10

MS240P0

 
thermo scientific™ lab vision™
MS240P0
200μg/mL 100µL Each for $238.04

MS240P1

 
thermo scientific™ lab vision™
MS240P1
200μg/mL 500µL Each for $409.63

MS240P1ABX

 
thermo scientific™ lab vision™
MS240P1ABX
1mg/mL 100µL Each for $538.67

MS240PABX

 
thermo scientific™ lab vision™
MS240PABX
1mg/mL 200µL Each for $687.49
Description & Specifications

Specifications

Antigen CD45/T200/LCA Ab-1
Clone Bra55
Applications Flow Cytometry
Applications Immunofluorescence
Applications Flow Cytometry
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Immunogen Non-T, non-B ALL cell line (REH)
Isotype IgG1

Ensure accurate, reproducible results in immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD45/T200/LCA Ab-1, Mouse Monoclonal Antibody.

CD45 leucocyte common antigen (LCA) belongs to the family of at least four isoforms of membrane glycoproteins (220, 205, 190, 180 kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and malignant non-hematopoietic tissues. The intracellular portion of these molecules have protein phosphatase activity and are involved in regulation of transmembrane signals.

Host Species: Mouse

Clone: Bra55

Isotype: IgG1

Species Reactivity: Human. Does not react with rhesus monkey. Others not tested.

Epitope: Not determined

Immunogen: Non-T, non-B ALL cell line (REH)

Molecular Weight: 220, 205, 190, 180 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO