Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD45/T200/LCA Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD45/T200/LCA Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS240P1ABX

 View more versions of this product

Catalog No. MS240P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen CD45/T200/LCA Ab-1
Applications Flow Cytometry
Applications Immunofluorescence
Classification Monoclonal
Clone Bra55
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Immunogen Non-T, non-B ALL cell line (REH)
Isotype IgG1
Quantity 100μL
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Ensure accurate, reproducible results in immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD45/T200/LCA Ab-1, Mouse Monoclonal Antibody.

CD45 leucocyte common antigen (LCA) belongs to the family of at least four isoforms of membrane glycoproteins (220, 205, 190, 180 kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and malignant non-hematopoietic tissues. The intracellular portion of these molecules have protein phosphatase activity and are involved in regulation of transmembrane signals.

Host Species: Mouse

Clone: Bra55

Isotype: IgG1

Species Reactivity: Human. Does not react with rhesus monkey. Others not tested.

Epitope: Not determined

Immunogen: Non-T, non-B ALL cell line (REH)

Molecular Weight: 220, 205, 190, 180 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO