Please login to your online account to display your discounted pricing

CD45 Rat anti-Mouse, PE-Cy7, Clone: 30-F11, eBioscience™

Rat Monoclonal Antibody

$172.55 - $259.55

Specifications

Antigen CD45
Clone 30-F11
Host Species Rat
Gene Alias Leukocyte Common Antigen, LCA, Ly-5
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015513

 
affymetrix
25045181
50μg Each for $172.55

501129643

 
affymetrix
25045182
100μg Each for $259.55
Description & Specifications

Specifications

Antigen CD45
Clone 30-F11
Host Species Rat
Gene Alias Leukocyte Common Antigen, LCA, Ly-5
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 30-F11 monoclonal antibody reacts with all isoforms of mouse CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding mature erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes.