Please login to your online account to display your discounted pricing

CD45 Rat anti-Mouse, PE, Clone: 30-F11, eBioscience™

Rat Monoclonal Antibody

$129.05 - $245.05

Specifications

Antigen CD45
Clone 30-F11
Host Species Rat
Gene Alias Leukocyte Common Antigen, LCA, Ly-5
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010370

 
affymetrix
12045182
100μg Each for $143.55

5010371

 
affymetrix
12045183
200μg Each for $245.05

501129438

 
affymetrix
12045181
50μg Each for $129.05
Description & Specifications

Specifications

Antigen CD45
Clone 30-F11
Host Species Rat
Gene Alias Leukocyte Common Antigen, LCA, Ly-5
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 30-F11 monoclonal antibody reacts with all isoforms of mouse CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding mature erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes.