missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD45 Rat anti-Mouse, FITC, Clone: 30-F11, eBioscience™

Rat Monoclonal Antibody

$71.05 - $375.55

Specifications

Antigen CD45
Clone 30-F11
Host Species Rat
Gene Alias Leukocyte Common Antigen, LCA, Ly-5
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509570 Life Technologies
LS11045181
50μg Each for $71.05
501129405 Life Technologies
LS11045182
100μg Each for $114.55
501129596 Life Technologies
LS11045185
500μg Each for $375.55
Specifications

Description

The 30-F11 monoclonal antibody reacts with all isoforms of mouse CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding mature erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes.
Description & Specifications

Specifications

CD45
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
30-F11
Leukocyte Common Antigen, LCA, Ly-5
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL