Please login to your online account to display your discounted pricing

CD45 Rat anti-Mouse, eVolve 605, Clone: 30-F11, eBioscience™

Rat Monoclonal Antibody

$210.25 - $427.75

Specifications

Antigen CD45
Clone 30-F11
Host Species Rat
Gene Alias Leukocyte Common Antigen, LCA, Ly-5
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125013

 
life technologies
LS83045141
25 Tests Each for $210.25

501125014

 
life technologies
LS83045142
100 Tests Each for $427.75
Description & Specifications

Specifications

Antigen CD45
Clone 30-F11
Host Species Rat
Gene Alias Leukocyte Common Antigen, LCA, Ly-5
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eVolve 605
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL

The 30-F11 monoclonal antibody reacts with all isoforms of mouse CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding mature erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes.