Please login to your online account to display your discounted pricing

CD45 Rat anti-Canine, FITC, Clone: YKIX716.13, eBioscience™

Rat Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen CD45
Clone YKIX716.13
Host Species Rat
Species Reactivity Canine
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509837

 
life technologies
LS11545041
25 Tests Each for $143.55

509838

 
life technologies
LS11545042
100 Tests Each for $361.05
Description & Specifications

Specifications

Antigen CD45
Clone YKIX716.13
Host Species Rat
Species Reactivity Canine
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

This YKIX716.13 monoclonal antibody reacts with canine CD45, also known as the leukocyte common antigen. CD45 is expressed by all hematopoietic cells except red blood cells and platelets. CD45 exists as multiple isoforms due to alternative splicing. The C-terminal domain of CD45 exhibits tyrosine phosphatase activity that enables signaling by T cell and B cell receptors following activation.

This monoclonal antibody recognizes all isoforms of CD45. In addition, when used in conjunction with YKIX753.22, this antibody has been reported to promote cell depletion via antibody-dependent cell-mediated cytotoxicity.