Please login to your online account to display your discounted pricing

CD45 Mouse anti-Rat, eFluor 450, Clone: OX1, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $332.05

Specifications

Antigen CD45
Clone OX1
Host Species Murine
Gene Alias Leukocyte Common Antigen
Species Reactivity Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124113

 
life technologies
LS48046180
25μg Each for $129.05

501124114

 
life technologies
LS48046182
100μg Each for $332.05
Description & Specifications

Specifications

Antigen CD45
Clone OX1
Host Species Murine
Gene Alias Leukocyte Common Antigen
Species Reactivity Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This OX1 monoclonal antibody recognizes a non-polymorphic determinant of rat CD45, which is also known as the leukocyte common antigen. CD45 is expressed on all hematopoietic cells except erythrocytes and platelets. This type I transmembrane protein tyrosine phosphatase exists as a variety of different isoforms due to alternative splicing.