missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD45 Mouse anti-Rat, APC, Clone: OX1, eBioscience™

Mouse Monoclonal Antibody

$117.48 - $302.28

Specifications

Antigen CD45
Clone OX1
Host Species Murine
Gene Alias Leukocyte Common Antigen
Species Reactivity Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245646 Life Technologies
LS17046180
25μg Each for $117.48
50245647 Life Technologies
LS17046182
100μg Each for $302.28
Specifications

Description

This OX1 monoclonal antibody recognizes a non-polymorphic determinant of rat CD45, which is also known as the leukocyte common antigen. CD45 is expressed on all hematopoietic cells except erythrocytes and platelets. This type I transmembrane protein tyrosine phosphatase exists as a variety of different isoforms due to alternative splicing.
Description & Specifications

Specifications

CD45
Murine
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
OX1
Leukocyte Common Antigen
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL