Please login to your online account to display your discounted pricing

CD45 Mouse anti-Human, PerCP-Cy5.5, Clone: 2D1, eBioscience™

Mouse Monoclonal Antibody

$220.40 - $516.20

Specifications

Antigen CD45
Clone 2D1
Host Species Mouse
Gene Alias LCA
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015884

 
life technologies
LS45945941
25 tests Each for $220.40

5015885

 
life technologies
LS45945942
100 tests Each for $516.20
Description & Specifications

Specifications

Antigen CD45
Clone 2D1
Host Species Mouse
Gene Alias LCA
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-Cy5.5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The 2D1 monoclonal antibody reacts with all isoforms of human CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding circulating erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes.