Please login to your online account to display your discounted pricing

CD45 Mouse anti-Human, PE-eFluor 610, Clone: HI30, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $448.05

Specifications

Antigen CD45
Clone HI30
Host Species Mouse
Gene Alias Leukocyte Common Antigen, LCA, Ly-5
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124766

 
life technologies
LS61045941
25 tests Each for $172.55

501124767

 
life technologies
LS61045942
100 tests Each for $448.05
Description & Specifications

Specifications

Antigen CD45
Clone HI30
Host Species Mouse
Gene Alias Leukocyte Common Antigen, LCA, Ly-5
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The HI30 monoclonal antibody reacts with all isoforms of human CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding circulating erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes.