missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD45 Mouse anti-Human, Biotin, Clone: HI30, eBioscience™

Mouse Monoclonal Antibody

$104.28 - $236.28

Specifications

Antigen CD45
Clone HI30
Host Species Murine
Gene Alias Leukocyte Common Antigen, LCA, Ly-5
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011569 Life Technologies
LS13045980
25μg Each for $104.28
501129428 Life Technologies
LS13045982
100μg Each for $236.28
Specifications

Description

The HI30 monoclonal antibody reacts with all isoforms of human CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding circulating erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes.
Description & Specifications

Specifications

CD45
Murine
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
HI30
Leukocyte Common Antigen, LCA, Ly-5
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer