Please login to your online account to display your discounted pricing

CD45 Mouse anti-Human, Biotin, Clone: CD45-2B11, eBioscience™

Mouse Monoclonal Antibody

$117.45 - $294.35

Specifications

Antigen CD45
Clone CD45-2B11
Host Species Murine
Gene Alias LCA
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122584

 
life technologies
LS13945780
25μg Each for $117.45

501122585

 
life technologies
LS13945782
100μg Each for $294.35
Description & Specifications

Specifications

Antigen CD45
Clone CD45-2B11
Host Species Murine
Gene Alias LCA
Species Reactivity Human
Applications Immunohistochemistry (Formalin/Paraffin)
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The CD45-2B11 monoclonal antibody reacts with human CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding circulating erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in lymphocyte proliferation and differentiation. The 2B11 antibody is useful for recognition of normal and neoplastic lymphoid cells.