missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD45.2, eFluor 506, clone: 104, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS69045482

 View more versions of this product

Catalog No. 501124950



Description

Description

CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing; these isoforms differ in their extracellular domains, whereas they share identical transmembrane and cytoplasmic domains. These isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. Besides the role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells.

CD45.2 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45.2 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45.2 protein exists as multiple isoforms as a result of alternative splicing; these isoforms differ in their extracellular domains, whereas they share identical transmembrane and cytoplasmic domains. These isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. Besides the role in immunoreceptor signaling, CD45.2 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells.
Specifications

Specifications

CD45.2
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P06800
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C, PTPRC
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
104
eFluor® 506
Ptprc
Liquid
Ptprc
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
19264
Documents

For Research Use Only.