missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD45.2 Mouse anti-Mouse, FITC, Clone: 104, eBioscience™

Mouse Monoclonal Antibody

$104.28 - $394.68

Specifications

Antigen CD45.2
Clone 104
Host Species Murine
Gene Alias C57BL/6, Balb/c, Ly5.2
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509581 Life Technologies
LS11045481
50μg Each for $104.28
509582 Life Technologies
LS11045482
100μg Each for $183.48
509583 Life Technologies
LS11045485
500μg Each for $394.68
Specifications

Description

The 104 monoclonal antibody reacts with the mouse CD45 molecule, the leukocyte common antigen (LCA) in CD45.2-expressing mouse strains. The strains that express CD45.2 include the most commonly used mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. CD45.2 is expressed by all leukocytes in these strains.
Description & Specifications

Specifications

CD45.2
Murine
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL
104
C57BL/6, Balb/c, Ly5.2
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer