Please login to your online account to display your discounted pricing

CD45.2 Mouse anti-Mouse, Biotin, Clone: 104, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $361.05

Specifications

Antigen CD45.2
Clone 104
Host Species Mouse
Gene Alias C57BL/6, Balb/c, Ly5.2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011558

 
life technologies
LS13045481
50μg Each for $114.55

5011559

 
life technologies
LS13045482
100μg Each for $216.05

5011560

 
life technologies
LS13045485
500μg Each for $361.05
Description & Specifications

Specifications

Antigen CD45.2
Clone 104
Host Species Mouse
Gene Alias C57BL/6, Balb/c, Ly5.2
Species Reactivity Mouse
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 104 monoclonal antibody reacts with the mouse CD45 molecule, the leukocyte common antigen (LCA) in CD45.2-expressing mouse strains. The strains that express CD45.2 include the most commonly used mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. CD45.2 is expressed by all leukocytes in these strains.