Please login to your online account to display your discounted pricing

CD45.2 Mouse anti-Mouse, APC-eFluor 780, Clone: 104, eBioscience™

Mouse Monoclonal Antibody

$176.90 - $413.25

Specifications

Antigen CD45.2
Clone 104
Host Species Mouse
Gene Alias C57BL/6, Balb/c, Ly5.2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124027

 
life technologies
LS47045480
25μg Each for $176.90

501129665

 
life technologies
LS47045482
100μg Each for $413.25
Description & Specifications

Specifications

Antigen CD45.2
Clone 104
Host Species Mouse
Gene Alias C57BL/6, Balb/c, Ly5.2
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC-eFluor 780
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 104 monoclonal antibody reacts with the mouse CD45 molecule, the leukocyte common antigen (LCA) in CD45.2-expressing mouse strains. The strains that express CD45.2 include the most commonly used mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. CD45.2 is expressed by all leukocytes in these strains.