Learn More
CD45.1 Monoclonal Antibody (A20), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen 48045382
Description
Description: The A20 monoclonal antibody reacts with the mouse CD45 molecule, the leukocyte common antigen (LCA) in CD45.1-expressing mouse strains. The strains that express CD45.1 include SJL/J, DA, STS/A and RIII. CD45.1 is expressed by all leukocytes in these strains. Applications Reported: This A20 antibody has been reported for use in flow cytometric analysis. Applications Tested: This A20 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD45.1 Monoclonal antibody specifically detects CD45.1 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD45.1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P06800 | |
Ptprc | |
Affinity chromatography | |
RUO | |
19264 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
A20 | |
eFluor 450 | |
Ptprc | |
B220; Cd45; CD45 antigen; CD45R; GP180; Lca; L-CA; Leukocyte common antigen; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
Mouse | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.