Please login to your online account to display your discounted pricing

CD45.1 Mouse anti-Mouse, Biotin, Clone: A20, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $361.05

Specifications

Antigen CD45.1
Clone A20
Host Species Murine
Gene Alias SJL, Ly5.1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011555

 
life technologies
LS13045381
50μg Each for $114.55

5011556

 
life technologies
LS13045382
100μg Each for $216.05

5011557

 
life technologies
LS13045385
500μg Each for $361.05
Description & Specifications

Specifications

Antigen CD45.1
Clone A20
Host Species Murine
Gene Alias SJL, Ly5.1
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The A20 monoclonal antibody reacts with the mouse CD45 molecule, the leukocyte common antigen (LCA) in CD45.1-expressing mouse strains. The strains that express CD45.1 include SJL/J, DA, STS/A and RIII. CD45.1 is expressed by all leukocytes in these strains.