Please login to your online account to display your discounted pricing

CD44H Mouse anti-Rat, PE, Clone: OX49, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen CD44H
Clone OX49
Host Species Mouse
Gene Alias CD44s, Pgp-1, H-CAM
Species Reactivity Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122233

 
life technologies
LS12044480
25μg Each for $143.55

501122234

 
life technologies
LS12044482
100μg Each for $361.05
Description & Specifications

Specifications

Antigen CD44H
Clone OX49
Host Species Mouse
Gene Alias CD44s, Pgp-1, H-CAM
Species Reactivity Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This OX49 monoclonal antibody recognizes rat CD44H, which is also known as homing-associated cell adhesion molecule (H-CAM) or phagocytic glycoprotein-1 (Pgp-1). This isoform of CD44 is the cell surface receptor for hyaluronate, a component of the extracellular matrix. CD44H is highly expressed on hematopoietic and non-hematopoietic cells, and exists as multiple splice variants. CD44H plays a critical role in lymphocyte activation and homing.