missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD44 Std./H-CAM Ab-2, Mouse Monoclonal Antibody

Ensure accurate and reproducible results in immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD44 Std./H-CAM Ab-2, Mouse Monoclonal Antibody.

$243.12 - $1,095.63

Specifications

Antigen CD44 Std./H-CAM Ab-2
Clone 5F12
Applications Flow Cytometry
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS178P Thermo Scientific™ Lab Vision™
MS178P
200μg/mL 1mL Each for $1,010.04
MS178P0 Thermo Scientific™ Lab Vision™
MS178P0
200μg/mL 100μL Each for $243.12
MS178P1 Thermo Scientific™ Lab Vision™
MS178P1
200μg/mL 500μL Each for $676.16
MS178P1ABX Thermo Scientific™ Lab Vision™
MS178P1ABX
1mg/mL 100μL Each for $775.45
MS178PABX Thermo Scientific™ Lab Vision™
MS178PABX
1mg/mL 200μL Each for $1,095.63
Specifications

Description

Ensure accurate and reproducible results in immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD44 Std./H-CAM Ab-2, Mouse Monoclonal Antibody.

CD44 is a cell surface glycoprotein expressed on lymphocytes, monocytes, and granulocytes. CD44 is also known as homing cell adhesion molecule (H-CAM), Phagocytic glycoprotein-1 (PgP-1), ECM-III, HUTCH-1, or Hermes-1. CD44 has been implicated in cell migration, in lymphocyte homing, and in tumor metastasis. Ligand for CD44 is hyaluronan of extracellular matrix. Ab-2 blocks the binding of hyaluronic acid to CD44 receptor.

Host Species: Mouse

Clone: 5F12

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Partially sensitive to trypsin

Immunogen: EBV-transformed normal human B-lymphocytes

Molecular Weight: 80-95 kDa

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence
  • Flow Cytometry
  • Blocks the binding of hyaluronic acid to CD44 receptor

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

CD44 Std./H-CAM Ab-2
Flow Cytometry
Unlabeled
Murine
Oncogenesis
EBV transformed normal human B-lymphocytes
5F12
Monoclonal
Concentrated
RUO
Primary
IgG1