Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD44 Std./H-CAM Ab-2, Mouse Monoclonal Antibody


Ensure accurate and reproducible results in immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD44 Std./H-CAM Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS178P1ABX

 View more versions of this product

Catalog No. MS178P1ABX

  • / Each
 

Description & Specifications

Specifications

Immunogen EBV transformed normal human B-lymphocytes
Quantity 100µL
Host Species Mouse
Regulatory Status RUO
Clone 5F12
Cross Reactivity Human
Format Concentrated
Applications Flow Cytometry
Isotype IgG1
Antigen CD44 Std./H-CAM Ab-2
Conjugate Unlabeled
Research Discipline Oncogenesis
Concentration 1mg/mL
Classification Monoclonal
Type Primary

Ensure accurate and reproducible results in immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD44 Std./H-CAM Ab-2, Mouse Monoclonal Antibody.

CD44 is a cell surface glycoprotein expressed on lymphocytes, monocytes, and granulocytes. CD44 is also known as homing cell adhesion molecule (H-CAM), Phagocytic glycoprotein-1 (PgP-1), ECM-III, HUTCH-1, or Hermes-1. CD44 has been implicated in cell migration, in lymphocyte homing, and in tumor metastasis. Ligand for CD44 is hyaluronan of extracellular matrix. Ab-2 blocks the binding of hyaluronic acid to CD44 receptor.

Host Species: Mouse

Clone: 5F12

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Partially sensitive to trypsin

Immunogen: EBV-transformed normal human B-lymphocytes

Molecular Weight: 80-95 kDa

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence
  • Flow Cytometry
  • Blocks the binding of hyaluronic acid to CD44 receptor

Note:

USA: RUO; Int'l: RUO