missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD44, PE-eFluor 610, clone: IM7, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS61044180

 View more versions of this product

Catalog No. 501124757



Description

Description

CD44 cell surface antigen is a 100kDa glycoprotein widely expressed on human leucocytes, white matter of the brain and by some epithelial cells of the intestine and breast. Several isoforms of CD44 exist, including the predominant CD44H isoform detected in many normal tissues. CD44 is a receptor for hyaluronic acid (HA) and is involved in cell-cell interactions, cell adhesion and migration. CD44 also participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing. CD44 expression may be up-regulated upon some carcinomas, and it has been speculated that this may be related to metastatic potential.

00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CD44 cell surface antigen is a 100 kDa type 1 transmembrane glycoprotein widely expressed on human leucocytes, white matter of the brain and by some epithelial cells of the intestine and breast. Several isoforms of CD44 exist, including the predomit CD44H isoform detected in many normal tissues. CD44 is a receptor for hyaluronic acid (HA) and is involved in cell-cell interactions, cell adhesion and migration. CD44 also participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing. CD44 expression may be up-regulated upon some carcinomas, and it has been speculated that this may be related to metastatic potential. CD44 is expressed by hematopoietic, non-hematopoietic cells, epithelial tissues, and to filopodia in cultured keratinocytes. Further, bone marrow myeloid cells and memory T cells express CD44 at high levels, and peripheral B and T cells can upregulate the expression of CD44 in response to certain stimulatory events. Transcripts for the CD44 gene undergo complex alternative splicing that results in many functionally distinct isoforms, however, the full-length nature of some of these variants have not been determined. Alternative splicing is the basis for the structural and functional diversity of the CD44 protein. Diseases associated with CD44 dysfunction include superficial keratitis and lichen sclerosus. CD44 also may be related to tumor metastasis formation.
Specifications

Specifications

CD44
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P16070, P15379
CD44 antigen, Cell surface glycoprotein CD44, hyaluronate binding protein, ECMR-III, GP90 lymphocyte homing adhesion receptor, HUTCH-I, PGP-1, extracellular matrix receptor III, hermes antigen, hyaluronate receptor, phagocytic glycoprotein 1, phagocytic glycoprotein I
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Flow Cytometry
IM7
PE-eFluor® 610
CD44
Liquid
CD44
IgG2b, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12505, 960
Documents

For Research Use Only.