Please login to your online account to display your discounted pricing

CD44 Rat anti-Human, Mouse, PE-Cy5, Clone: IM7, eBioscience™

Rat Monoclonal Antibody

$201.55 - $506.05

Specifications

Antigen CD44
Clone IM7
Host Species Rat
Gene Alias Pgp-1, MDU3, Hermes, Hyaluronate receptor
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014050

 
affymetrix
15044182
100μg Each for $274.05

5014049

 
affymetrix
15044181
50μg Each for $201.55

5014051

 
affymetrix
15044183
200μg Each for $506.05
Description & Specifications

Specifications

Antigen CD44
Clone IM7
Host Species Rat
Gene Alias Pgp-1, MDU3, Hermes, Hyaluronate receptor
Species Reactivity Human, Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The IM7 monoclonal antibody reacts with all isoforms of mouse CD44 (Pgp-1). CD44 is expressed by hematopoietic and non-hematopoietic cells. Bone marrow myeloid cells and memory T cells highly express this antigen and peripheral B and T cells can upregulate the expression of CD44. CD44 functions as an adhesion molecule through its binding to hyaluronate, an extracellular matrix component.