missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD44 Rat anti-Human, Mouse, FITC, Clone: IM7, eBioscience™

Rat Monoclonal Antibody

$100.05 - $665.55

Specifications

Antigen CD44
Clone IM7
Host Species Rat
Gene Alias Pgp-1, MDU3, Hermes, Hyaluronate receptor
Species Reactivity Human, Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509565 Life Technologies
LS11044181
50μg Each for $100.05
509566 Life Technologies
LS11044182
100μg Each for $187.05
509567 Life Technologies
LS11044185
500μg Each for $375.55
509568 Life Technologies
LS11044186
1mg Each for $665.55
Specifications

Description

The IM7 monoclonal antibody reacts with all isoforms of mouse CD44 (Pgp-1). CD44 is expressed by hematopoietic and non-hematopoietic cells. Bone marrow myeloid cells and memory T cells highly express this antigen and peripheral B and T cells can upregulate the expression of CD44. CD44 functions as an adhesion molecule through its binding to hyaluronate, an extracellular matrix component.
Description & Specifications

Specifications

CD44
Rat
Human, Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
IM7
Pgp-1, MDU3, Hermes, Hyaluronate receptor
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL