Please login to your online account to display your discounted pricing

CD44 Rat anti-Human, Mouse, Clone: IM7, eBioscience™

Rat Monoclonal Antibody

$85.55 - $506.05

Specifications

Antigen CD44
Clone IM7
Host Species Rat
Gene Alias Pgp-1, MDU3, Hermes, Hyaluronate receptor
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012579

 
life technologies
LS14044181
50μg Each for $85.55

5012580

 
life technologies
LS14044182
100μg Each for $158.05

5012581

 
life technologies
LS14044185
500μg Each for $303.05

5012582

 
life technologies
LS14044186
1mg Each for $506.05
Description & Specifications

Specifications

Antigen CD44
Clone IM7
Host Species Rat
Gene Alias Pgp-1, MDU3, Hermes, Hyaluronate receptor
Species Reactivity Human, Mouse
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The IM7 monoclonal antibody reacts with all isoforms of mouse CD44 (Pgp-1). CD44 is expressed by hematopoietic and non-hematopoietic cells. Bone marrow myeloid cells and memory T cells highly express this antigen and peripheral B and T cells can upregulate the expression of CD44. CD44 functions as an adhesion molecule through its binding to hyaluronate, an extracellular matrix component.