Please login to your online account to display your discounted pricing

CD44 Rat anti-Human, Mouse, APC-eFluor 780, Clone: IM7, eBioscience™

Rat Monoclonal Antibody

$162.40 - $427.75

Specifications

Antigen CD44
Clone IM7
Host Species Rat
Gene Alias Pgp-1, MDU3, Hermes, Hyaluronate receptor
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016163

 
life technologies
LS47044180
25μg Each for $162.40

501129456

 
life technologies
LS47044182
100μg Each for $427.75
Description & Specifications

Specifications

Antigen CD44
Clone IM7
Host Species Rat
Gene Alias Pgp-1, MDU3, Hermes, Hyaluronate receptor
Species Reactivity Human, Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC-eFluor 780
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The IM7 monoclonal antibody reacts with all isoforms of mouse CD44 (Pgp-1). CD44 is expressed by hematopoietic and non-hematopoietic cells. Bone marrow myeloid cells and memory T cells highly express this antigen and peripheral B and T cells can upregulate the expression of CD44. CD44 functions as an adhesion molecule through its binding to hyaluronate, an extracellular matrix component.