Please login to your online account to display your discounted pricing

CD44 Rat anti-Canine, FITC, Clone: YKIX337.8, eBioscience™

Rat Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen CD44
Clone YKIX337.8
Host Species Rat
Gene Alias Hyaluronate receptor
Species Reactivity Canine
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509833

 
life technologies
LS11544041
25 tests Each for $143.55

509834

 
life technologies
LS11544042
100 tests Each for $361.05
Description & Specifications

Specifications

Antigen CD44
Clone YKIX337.8
Host Species Rat
Gene Alias Hyaluronate receptor
Species Reactivity Canine
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This YKIX337.8 monoclonal antibody reacts with canine CD44, also known as phagocytic glycoprotein-1 (Pgp-1). CD44 is an adhesion molecule that binds hyaluronan, a major component of the extracellular matrix. This cell surface molecule is expressed on leukocytes and epithelial cells, as well as in canine mammary and melanocytic tumors. Furthermore, CD44 expression is induced by T cell activation.