missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD43/T-Cell Marker Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Thermo Scientific™ CD43/T-Cell Marker Ab-3, Mouse Monoclonal Antibody.

$245.55 - $772.20

Specifications

Antigen CD43/T-Cell Marker Ab-3
Clone 84-3C1
Applications Flow Cytometry,Immunohistochemistry (Paraffin),Flow Cytometry,Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS679P Thermo Scientific™ Lab Vision™
MS679P
Purified Ab with BSA and Azide 1mL Each for $682.29
MS679P0 Thermo Scientific™ Lab Vision™
MS679P0
Purified Ab with BSA and Azide 100μL Each for $245.55
MS679P1 Thermo Scientific™ Lab Vision™
MS679P1
Purified Ab with BSA and Azide 500μL Each for $479.39
MS679P1ABX Thermo Scientific™ Lab Vision™
MS679P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $609.59
MS679PABX Thermo Scientific™ Lab Vision™
MS679PABX
Ab without BSA/Azide (Frozen) 200μL Each for $772.20
MS679R7 Thermo Scientific™ Lab Vision™
MS679R7
Ready-to-use for IHC (UVLP) 7mL Each for $352.37
Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Thermo Scientific™ CD43/T-Cell Marker Ab-3, Mouse Monoclonal Antibody.

CD43 (leukosialin, sialophorin, or leukocyte sialoglycoprotein) is a cell surface glycoprotein which is expressed on all thymocytes and T-cells. CD43 is involved in activation of T cells, B cells, NK cells, and monocytes

Host Species: Mouse

Clone: 84-3C1

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not tested.

Epitope: Not determined

Immunogen: Stimulated human leukocytes

Molecular Weight: 95/115/135 kDa (depending upon extent of glycosylation)

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

CD43/T-Cell Marker Ab-3
Flow Cytometry,Immunohistochemistry (Paraffin),Flow Cytometry,Immunohistochemistry (Paraffin)
Unlabeled
RUO
Primary
IgG1
84-3C1
Monoclonal
Murine
T-Cell
Stimulated human leukocytes