missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD43/T-Cell Marker Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Thermo Scientific™ CD43/T-Cell Marker Ab-3, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS679P1ABX

 View more versions of this product

Catalog No. MS679P1ABX


 

Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Thermo Scientific™ CD43/T-Cell Marker Ab-3, Mouse Monoclonal Antibody.

CD43 (leukosialin, sialophorin, or leukocyte sialoglycoprotein) is a cell surface glycoprotein which is expressed on all thymocytes and T-cells. CD43 is involved in activation of T cells, B cells, NK cells, and monocytes

Host Species: Mouse

Clone: 84-3C1

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not tested.

Epitope: Not determined

Immunogen: Stimulated human leukocytes

Molecular Weight: 95/115/135 kDa (depending upon extent of glycosylation)

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

CD43/T-Cell Marker Ab-3
Monoclonal
1mg/mL
Ab without BSA/Azide (Frozen)
Murine
IgG1
RUO
Primary
Human
Flow Cytometry,Immunohistochemistry (Paraffin)
84-3C1
Unlabeled
Concentrated
Stimulated human leukocytes
100μL
T-Cell
Monoclonal