missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible reults in immunohistochemistry and flow cytometry applications with Thermo Scientific™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody.

$318.53 - $631.65

Specifications

Antigen CD43/T-Cell Marker Ab-1
Clone DF-T1
Applications Flow Cytometry,Immunohistochemistry (Paraffin),Flow Cytometry,Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS146P Thermo Scientific™ Lab Vision™
MS146P
Purified Ab with BSA and Azide 1mL Each for $565.69
MS146P1 Thermo Scientific™ Lab Vision™
MS146P1
Purified Ab with BSA and Azide 500μL Each for $386.37
MS146P1ABX Thermo Scientific™ Lab Vision™
MS146P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $486.35
MS146PABX Thermo Scientific™ Lab Vision™
MS146PABX
Ab without BSA/Azide (Frozen) 200μL Each for $631.65
MS146R7 Thermo Scientific™ Lab Vision™
MS146R7
Ready-to-use for IHC (UVLP) 7mL Each for $318.53
Specifications

Description

Ensure accurate, reproducible reults in immunohistochemistry and flow cytometry applications with Thermo Scientific™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody.

CD43 (leukosialin, sialophorin, or leukocyte sialoglycoprotein) is a cell surface glycoprotein which is expressed on all thymocytes and T-cells. CD43 is involved in activation of T cells, B cells, NK cells, and monocytes

Host Species: Mouse

Clone: DF-T1

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not tested

Epitope: Not determined

Immunogen: Myeloblastic KG1 cells

Molecular Weight: 95/115/135 kDa (depending upon extent of glycosylation)

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry
Description & Specifications

Specifications

CD43/T-Cell Marker Ab-1
Flow Cytometry,Immunohistochemistry (Paraffin),Flow Cytometry,Immunohistochemistry (Paraffin)
Unlabeled
IVD
Primary
DF-T1
Monoclonal
Murine
T-Cell
Myeloblastic KG1 cells