Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible reults in immunohistochemistry and flow cytometry applications with Thermo Scientific™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS146P1ABX

 View more versions of this product

Catalog No. MS146P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen CD43/T-Cell Marker Ab-1
Host Species Mouse
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Regulatory Status IVD
Isotype IgG1
Clone DF-T1
Cross Reactivity Human
Format Concentrated
Quantity 100µL
Immunogen Myeloblastic KG1 cells
Classification Monoclonal
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)
Research Discipline T-Cell
Type Primary
Concentration 1mg/mL
Item Description 100uL; 1mg/mL; Purified w/o BSA, azide

Ensure accurate, reproducible reults in immunohistochemistry and flow cytometry applications with Thermo Scientific™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody.

CD43 (leukosialin, sialophorin, or leukocyte sialoglycoprotein) is a cell surface glycoprotein which is expressed on all thymocytes and T-cells. CD43 is involved in activation of T cells, B cells, NK cells, and monocytes

Host Species: Mouse

Clone: DF-T1

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not tested

Epitope: Not determined

Immunogen: Myeloblastic KG1 cells

Molecular Weight: 95/115/135 kDa (depending upon extent of glycosylation)

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry