Please login to your online account to display your discounted pricing

CD43 Mouse anti-Human, FITC, Clone: eBio84-3C1 (84-3C1), eBioscience™

Mouse Monoclonal Antibody

$114.55 - $259.55

Specifications

Antigen CD43
Clone eBio84-3C1 (84-3C1)
Host Species Murine
Gene Alias Leukosialin, LSN, Sialophorin, SPN
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245236

 
life technologies
LS11043941
25 Tests Each for $114.55

509563

 
life technologies
LS11043942
100 tests Each for $259.55
Description & Specifications

Specifications

Antigen CD43
Clone eBio84-3C1 (84-3C1)
Host Species Murine
Gene Alias Leukosialin, LSN, Sialophorin, SPN
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The monoclonal antibody ebio84-3C1 reacts with CD43, which is also known as leukosialin, galactoglycoprotein and sialophorin. CD43 is a sialomucin which like many mucins can have both adhesive and anti-adhesive functions. Expression of CD43 is found on most leukocytes except resting B lymphocytes. Proteolytic processing upon activation decreases surface expression. CD43 is involved in activation of T cells, B cells, NK cells, and monocytes. The counter-receptor for CD43 is CD169/SIGLEC-1, which is expressed on macrophages.