missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD42d Armenian Hamster anti-Mouse, Rat, PE, Clone: 1C2, eBioscience™

Armenian Hamster Monoclonal Antibody

$130.68 - $236.28

Specifications

Antigen CD42d
Clone 1C2
Host Species Armenian Hamster
Gene Alias platelet glycoprotein V, GP5, GPV
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245267 Life Technologies
LS12042180
25μg Each for $130.68
50245268 Life Technologies
LS12042182
100μg Each for $236.28
Specifications

Description

The monoclonal antibody 1C2 recognizes mouse and rat CD42d, a single transmembrane protein also known as platelet glycoprotein V (GPV). CD42d interacts with CD42a, CD42b and CD42c to form the receptor for von willebrand factor and thrombin. This functional complex plays a role in platelet adhesion. Expression of CD42d (unlike other markers such as CD41) is restricted to resting platelets and megakaryocytes and therefore is a good marker for development of megakaryocytes.
Description & Specifications

Specifications

CD42d
Armenian Hamster
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
1C2
platelet glycoprotein V, GP5, GPV
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer