Please login to your online account to display your discounted pricing

CD42b Mouse anti-Human, FITC, Clone: HIP1, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $307.40

Specifications

Antigen CD42b
Clone HIP1
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122062

 
life technologies
LS11042941
25 Tests Each for $129.05

501122063

 
life technologies
LS11042942
100 Tests Each for $307.40
Description & Specifications

Specifications

Antigen CD42b
Clone HIP1
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1μg)/test

The HIP1 monoclonal antibody reacts with the human CD42b molecule, a 135kDa transmembrane protein also known as platelet gpIb.