Please login to your online account to display your discounted pricing

CD42b Mouse anti-Human, Clone: HIP1, eBioscience™

Mouse Monoclonal Antibody

$71.05 - $158.05

Specifications

Antigen CD42b
Clone HIP1
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012572

 
affymetrix
14042980
25μg Each for $71.05

5012573

 
affymetrix
14042982
100μg Each for $158.05
Description & Specifications

Specifications

Antigen CD42b
Clone HIP1
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The HIP1 monoclonal antibody reacts with the human CD42b molecule, a 135kDa transmembrane protein also known as platelet gpIb.