Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD42b (GPIb) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD42b (GPIb) Ab-1, Mouse Monoclonal Antibody.

$281.84 - $1,426.33

Specifications

Antigen CD42b (GPIb) Ab-1
Clone 42C01
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1174P

 
thermo scientific™ lab vision™
MS1174P
Purified Ab with BSA and Azide 1mL Each for $1,321.73

MS1174P0

 
thermo scientific™ lab vision™
MS1174P0
Purified Ab with BSA and Azide 100µL Each for $281.84

MS1174P1

 
thermo scientific™ lab vision™
MS1174P1
Purified Ab with BSA and Azide 500µL Each for $783.63

MS1174P1ABX

 
thermo scientific™ lab vision™
MS1174P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $889.00

MS1174PABX

 
thermo scientific™ lab vision™
MS1174PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,426.33

MS1174R7

 
thermo scientific
MS1174R7
Ready-to-use for IHC (UVLP) 7mL Each for $693.63
Description & Specifications

Specifications

Antigen CD42b (GPIb) Ab-1
Clone 42C01
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Hematopoietic
Type Primary
Immunogen Recombinant CD42b
Isotype IgG1

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD42b (GPIb) Ab-1, Mouse Monoclonal Antibody.

The CD42b glycoprotein, also known as GPIb, is a cofactor of ristocetin-induced aggregation and is involved in the binding of platelets to blood vessel walls. The CD42b antigen is expressed on platelets and on megakaryocytes in bone marrow. The absence of CD42b antigen on platelets may indicate Bernard-Soulier disease.

Host Species: Mouse

Clone: 42C01

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Immunogen: Recombinant CD42b.

Molecular Weight: 135-145 kDa (α-chain) and 22-25 kDa (β-chain)

Positive Control: Tonsil

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO