Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD42b (GPIb) Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD42b (GPIb) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1174PABX

 View more versions of this product

Catalog No. MS1174PABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Immunogen Recombinant CD42b
Host Species Mouse
Clone 42C01
Isotype IgG1
Antigen CD42b (GPIb) Ab-1
Quantity 200µL
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Format Concentrated
Research Discipline Hematopoietic
Formulation Ab without BSA/Azide (Frozen)
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Item Description 200uL; 1mg/mL; Purified w/o BSA, azide

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD42b (GPIb) Ab-1, Mouse Monoclonal Antibody.

The CD42b glycoprotein, also known as GPIb, is a cofactor of ristocetin-induced aggregation and is involved in the binding of platelets to blood vessel walls. The CD42b antigen is expressed on platelets and on megakaryocytes in bone marrow. The absence of CD42b antigen on platelets may indicate Bernard-Soulier disease.

Host Species: Mouse

Clone: 42C01

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Immunogen: Recombinant CD42b.

Molecular Weight: 135-145 kDa (α-chain) and 22-25 kDa (β-chain)

Positive Control: Tonsil

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO