missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD41a Mouse anti-Human, PerCP-eFluor 710, Clone: HIP8, eBioscience™

Mouse Monoclonal Antibody

$195.75 - $384.25

Specifications

Antigen CD41a
Clone HIP8
Host Species Murine
Gene Alias Integrin alpha Iib, platelet GPIIb
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123755 Life Technologies
LS46041941
25 Tests Each for $195.75
501130748 Life Technologies
LS46041942
100 Tests Each for $384.25
Specifications

Description

The HIP8 monoclonal antibody reacts with the human CD41 molecule, the integrin alpha IIb also known as platelet GPIIb. CD41 non-covalently associates with integrin beta3 (GPIIIa, CD61) and is expressed by megakaryocytes and platelets. The CD41/CD61 complex is a receptor for fibronectin, fibrinogen, von Willebrand factor, vitronectin and thrombospondin and mediates platelets aggregation. HIP8 blocks platelet aggregation.
Description & Specifications

Specifications

CD41a
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
HIP8
Integrin alpha Iib, platelet GPIIb
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test