Please login to your online account to display your discounted pricing

CD41a Mouse anti-Human, FITC, Clone: HIP8, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $288.55

Specifications

Antigen CD41a
Clone HIP8
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509556

 
affymetrix
11041941
25 tests Each for $114.55

509557

 
affymetrix
11041942
100 tests Each for $288.55
Description & Specifications

Specifications

Antigen CD41a
Clone HIP8
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The HIP8 monoclonal antibody reacts with the human CD41 molecule, the integrin αIIb also known as platelet GPIIb. CD41 non-covalently associates with integrin β3 (GPIIIa, CD61) and is expressed by megakaryocytes and platelets. The CD41/CD61 complex is a receptor for fibronectin, fibrinogen, von Willebrand factor, vitronectin and thrombospondin and mediates platelets aggregation. HIP8 blocks platelet aggregation.