Please login to your online account to display your discounted pricing

CD41 Rat anti-Mouse, eFluor 450, Clone: eBioMWReg30 (MWReg30), eBioscience™

Rat Monoclonal Antibody

$205.90 - $413.25

Specifications

Antigen CD41
Clone eBioMWReg30 (MWReg30)
Host Species Rat
Gene Alias fibrinogen receptor, gpIIb/IIIa, integrin alpha Iib, ITGA2b
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016288

 
life technologies
LS48041180
25μg Each for $205.90

5016289

 
life technologies
LS48041182
100μg Each for $413.25
Description & Specifications

Specifications

Antigen CD41
Clone eBioMWReg30 (MWReg30)
Host Species Rat
Gene Alias fibrinogen receptor, gpIIb/IIIa, integrin alpha Iib, ITGA2b
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBioMWReg30 monoclonal antibody reacts with mouse CD41 (fibrinogen receptor, gpIIb, integrin alpha IIb). While initially thought to be expressed exclusively on the surface of platelets and megakaryocytes, it has been demonstrated that CD41 is also expressed on hematopoietic progenitors in the embryo, fetus and adult. CD41 associates with CD61 (gpIIIa, integrin beta III) to form a receptor which plays a major role in platelet function, including binding of several adhesion molecules such as fibrinogen, fibronectin and vitronectin.

Recently, the SLAM-family markers, CD48 and CD150 have been used to reliably identify hematopoietic stem cells (HSC). Specifically, it was found that CD150+CD48- bone marrow cells were highly efficient in their ability to confer long-term multi-lineage reconstitution in irradiated mice. Furthermore, the efficiency of reconstitution was enhanced when HSCs were further enriched through the exclusion of CD41+ cells. Thus, the use of CD150+CD48-CD41- as an expression profile efficiently identifies hematopoietic stem cells.