Please login to your online account to display your discounted pricing

CD40 Rat anti-Mouse, PerCP-eFluor 710, Clone: 1C10, eBioscience™

Rat Monoclonal Antibody

$191.40 - $471.25

Specifications

Antigen CD40
Clone 1C10
Host Species Rat
Gene Alias TNFRSF5
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501128974

 
life technologies
LS46040182
100μg Each for $471.25

501130747

 
life technologies
LS46040180
25μg Each for $191.40
Description & Specifications

Specifications

Antigen CD40
Clone 1C10
Host Species Rat
Gene Alias TNFRSF5
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 1C10 monoclonal antibody reacts with mouse CD40, a 45-50kDa type I transmembrane glycoprotein. CD40 is a member of the TNFR family and is expressed by mouse B lymphocytes, follicular dendritic cells, thymic epithelium, and a subset of peripheral T cells. CD40 regulates B cell development/maturation by inducing Ig isotype switching and in combination with other signals such as IL-4, protects B cells from surface Ig-induced apoptosis and promotes proliferation. Interaction of CD40 with CD154 (gp39), its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation.

The monoclonal antibody 1C10 is reported to have agonistic activity in vitro and in vivo.