missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD40 Rat anti-Mouse, PE, Clone: 1C10, eBioscience™

Rat Monoclonal Antibody

$143.55 - $404.55

Specifications

Antigen CD40
Clone 1C10
Host Species Rat
Gene Alias TNFRSF5
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010349 Life Technologies
LS12040181
50μg Each for $143.55
5010350 Life Technologies
LS12040182
100μg Each for $361.05
5010351 Life Technologies
LS12040183
200μg Each for $404.55
Specifications

Description

The 1C10 monoclonal antibody reacts with mouse CD40, a 45-50kDa type I transmembrane glycoprotein. CD40 is a member of the TNFR family and is expressed by mouse B lymphocytes, follicular dendritic cells, thymic epithelium, and a subset of peripheral T cells. CD40 regulates B cell development/maturation by inducing Ig isotype switching and in combination with other signals such as IL-4, protects B cells from surface Ig-induced apoptosis and promotes proliferation. Interaction of CD40 with CD154 (gp39), its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation.

The monoclonal antibody 1C10 is reported to be agonistic in vitro and in vivo.
Description & Specifications

Specifications

CD40
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
1C10
TNFRSF5
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL